Fluvastatin sodium salt

Supplier: Apollo Scientific

Synonyms: XU 62-320, rel-(3S,5R,E)-7-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoic acid sodium salt, Sodium rel-(3R,5S,E)-7-(3-(4-fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoate

BISN0143-50MG BISN0143-100MG
APOSBISN0143-50MGEA 112 GBP
APOSBISN0143-50MG APOSBISN0143-100MG
Fluvastatin sodium salt
Fluvastatin sodium salt

Fluvastatin Sodium is a synthetic indole-heptanoic-acid-derived inhibitor of hepatic HMG-CoA reductase with IC50 of 8 nM.

Formula: C₂₄H₂₅FNNaO₄
MW: 433,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00929076
CAS Number: 93957-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR