BIBF-0775 97%

Supplier: Apollo Scientific

Synonyms: (Z)-N-Ethyl-N-methyl-2-oxo-3-(phenyl((4-(piperidin-1-ylmethyl)phenyl)amino)methylene)indoline-6-carboxamide, BIBF0775

BISN0074-5MG BISN0074-10MG BISN0074-50MG
APOSBISN0074-5MGEA 231 GBP
APOSBISN0074-5MG APOSBISN0074-10MG APOSBISN0074-50MG
BIBF-0775 97%
BIBF-0775

BIBF0775 is an inhibitor of transforming growth factor β receptor I (TGFβRI) by soaking into the kinase domain of TGFβRI.

Formula: C₃₁H₃₄N₃₄O₂
MW: 494,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25976645
CAS Number: 334951-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR