HOE-140 98%
Supplier: Apollo Scientific
Synonyms:
Icatibant, ((2S,3aS,7aS)-1-((R)-2-(((S)-2-(2-((2S,4R)-1-(D-Arginyl-L-arginyl-L-prolyl)-4-hydroxypyrrolidine-2-carboxamido)acetamido)-3-(thiophen-2-yl)propanoyl)-L-seryl)-1,2,3,4-tetrahydroisoquinoline-3-carbonyl)octahydro-1H-indole-2-carbonyl)-L-arginine
BISN0006-1MG
BISN0006-5MG
APOSBISN0006-1MGEA
296
GBP
APOSBISN0006-1MG
APOSBISN0006-5MG
HOE-140 98%
HOE-140
Icatibant is a selective and specific antagonist of bradykinin B2 receptor with IC50 and Ki of 1.07 nM and 0.798 nM respectively.
Formula:
C₅₉H₈₉N₁₉O₁₃S MW: 1304,5 g/mol Storage Temperature: Ambient |
CAS Number:
130308-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...