Syringaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde

BIS1001-100G BIS1001-25G BIS1001-500G
APOSBIS1001-100GEA 95.7 GBP
APOSBIS1001-100G APOSBIS1001-25G APOSBIS1001-500G
Syringaldehyde 98%
Syringaldehyde
Formula: C₉H₁₀O₄
MW: 182,18 g/mol
Boiling Pt: 192…193 °C/14mmHg
Melting Pt: 110…113 °C
Density: 1,013 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006943
CAS Number: 134-96-3
EINECS: 205-167-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR