2-[2-(2-Aminoethoxy)ethoxy]acetic acid 99%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(2-(2-Aminoethoxy)ethoxy)acetic acid, H2N-PEG2-CH2COOH, 8-Amino-3,6-dioxaoctanoic acid, Amino-PEG2-acetic acid

BIPG1063-1G BIPG1063-5G
APOSBIPG1063-1GEA 154 GBP
APOSBIPG1063-1G APOSBIPG1063-5G
2-[2-(2-Aminoethoxy)ethoxy]acetic acid 99%
2-[2-(2-Aminoethoxy)ethoxy]acetic acid
Formula: C₆H₁₃NO₄
MW: 163,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185897
CAS Number: 134978-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR