(4-Nitrophenyl)-β-D-glucopyranoside
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
p-Nitrophenyl-β-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
BIM1207-10G
BIM1207-5G
BIM1207-2G
APOSBIM1207-10GEA
387
GBP
APOSBIM1207-10G
APOSBIM1207-5G
APOSBIM1207-2G
(4-Nitrophenyl)-β-D-glucopyranoside
(4-Nitrophenyl) β-D-glucopyranoside
Substrate for b-Glucosidase
Formula:
C₁₂H₁₅NO₈ MW: 301,25 g/mol Melting Pt: 166…167 °C Storage Temperature: Freezer |
MDL Number:
MFCD00006593 CAS Number: 2492-87-7 EINECS: 219-661-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...