4-Methylumbelliferyl-Β-D-cellobiopyranoside

Supplier: Apollo Scientific
Warning

Synonyms: 4-Methylumbelliferyl beta-D-Cellobioside

BIM1091-100MG BIM1091-500MG
APOSBIM1091-100MGEA 52.5 GBP
APOSBIM1091-100MG APOSBIM1091-500MG
4-Methylumbelliferyl-Β-D-cellobiopyranoside
4-Methylumbelliferyl-Β-D-cellobiopyranoside
Formula: C₂₂H₂₈O₁₃
MW: 500,45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063279
CAS Number: 72626-61-0
EINECS: 276-744-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR