Minoxidil
Supplier: Apollo Scientific
|
Warning
|
BIM0127-1G
BIM0127-25G
BIM0127-5G
APOSBIM0127-1GEA
19.7
GBP
APOSBIM0127-1G
APOSBIM0127-25G
APOSBIM0127-5G
Minoxidil
Minoxidil
Formula:
C₉H₁₅N₅O Melting Pt: 272…274 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063409 CAS Number: 38304-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...