Lovastatin

Supplier: Apollo Scientific
Warning

Synonyms: 6alpha-Methylcompactin, Monacolin K, Mevinolin, (S)-(1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2-methylbutanoate

BIL1008-100MG BIL1008-25MG BIL1008-50MG
APOSBIL1008-100MGEA 393 GBP
APOSBIL1008-100MG APOSBIL1008-25MG APOSBIL1008-50MG
Lovastatin
Lovastatin
Formula: C₂₄H₃₆O₅
MW: 404,55 g/mol
Melting Pt: 175 °C
Storage Temperature: Freezer
MDL Number: MFCD00072164
CAS Number: 75330-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR