α-Hydroxy-o-tolyl-β-D-glucopyranoside

Supplier: Apollo Scientific

Synonyms: 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin

BIK9020-10MG BIK9020-25MG BIK9020-100MG
APOSBIK9020-10MGEA 21.1 GBP
APOSBIK9020-10MG APOSBIK9020-25MG APOSBIK9020-100MG
α-Hydroxy-o-tolyl-β-D-glucopyranoside
α-Hydroxy-o-tolyl β-D-glucopyranoside
Formula: C₁₃H₁₈O₇
MW: 286,28 g/mol
Melting Pt: 196…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00006590
CAS Number: 138-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR