2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside 99% min

Supplier: Apollo Scientific
Warning

Synonyms: 2-Azidoethyl 2,3,4,6-Tetra-O-acetyl-β-D-glucopyranoside

BICL4024-1G BICL4024-2G BICL4024-500MG
APOSBICL4024-1GEA 453 GBP
APOSBICL4024-1G APOSBICL4024-2G APOSBICL4024-500MG
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside 99% min
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranoside
Formula: C₁₆H₂₃N₃O₁₀
Melting Pt: 114…115 °C
Storage Temperature: Freezer
CAS Number: 140428-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR