Cytochalasin E

Supplier: Apollo Scientific
Danger

BIC1017-10MG BIC1017-1MG BIC1017-5MG
APOSBIC1017-10MGEA 655 GBP
APOSBIC1017-10MG APOSBIC1017-1MG APOSBIC1017-5MG
Cytochalasin E
Cytochalasin E
Formula: C₂₈H₃₃NO₇
Melting Pt: 206 °C
Storage Temperature: Freezer
CAS Number: 36011-19-5
EINECS: 252-835-7
UN: 1544
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR