1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
alpha,beta-D-Mannose pentaacetate, 1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, (3S,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate
BIB6096-100G
BIB6096-50G
APOSBIB6096-100GEA
385
GBP
APOSBIB6096-100G
APOSBIB6096-50G
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose
Formula:
C₁₆H₂₂O₁₁ MW: 390,34 g/mol Melting Pt: 68 °C Storage Temperature: Freezer |
MDL Number:
MFCD05864874 CAS Number: 25941-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...