5-Bromo-4-chloro-3-indoxyl-N-acetyl-beta-D-glucosaminide

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-4-chloro-3-indoxyl-2-acetamido-2-deoxy-beta-D-glucopyranoside, X-beta-D-GlcNAc

BIB1175-1G BIB1175-500MG BIB1175-5G
APOSBIB1175-1GEA 98.3 GBP
APOSBIB1175-1G APOSBIB1175-500MG APOSBIB1175-5G
5-Bromo-4-chloro-3-indoxyl-N-acetyl-beta-D-glucosaminide
5-Bromo-4-chloro-3-indoxyl-N-acetyl-beta-D-glucosaminide
Formula: C₁₆H₁₈BrClN₂O₆
MW: 449,69 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00040631
CAS Number: 4264-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR