TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
Supplier: Apollo Scientific
Synonyms:
Tris(hydroxymethyl)aminomethane hydrochloride
, Tris(hydroxymethyl)methylamine hydrochloride
, Trizma(R) hydrochloride
, Tris(hydroxymethyl)methylammonium chloride
, THAM HCl
, Trometamol hydrochloride
, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride
, TRIS HCl
APOSBIA3452-500GEA
165
GBP
APOSBIA3452-500G
APOSBIA3452-250G
APOSBIA3452-1KG
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
Formula:
NH₂C(CH₂OH)₃·HCl MW: 157,6 g/mol Melting Pt: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012590 CAS Number: 1185-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...