Amylose

Supplier: Apollo Scientific
Warning

BIA1408-10G BIA1408-100G BIA1408-250G
APOSBIA1408-10GEA 59 GBP
APOSBIA1408-10G APOSBIA1408-100G APOSBIA1408-250G
Amylose
Amylose

A polymer of D-glucose units of variable molecular weight.

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00134652
CAS Number: 9005-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR