Acyclovir

Supplier: Apollo Scientific
Warning

Synonyms: 1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on, Acycloguanosine, 2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one

BIA1250-100MG BIA1250-50MG
APOSBIA1250-100MGEA 46 GBP
APOSBIA1250-100MG APOSBIA1250-50MG
Acyclovir
Acyclovir
Formula: C₈H₁₁N₅O₃
MW: 225,21 g/mol
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00057880
CAS Number: 59277-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR