Azithromycin
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one
BIA0165-1G
BIA0165-5G
BIA0165-10G
APOSBIA0165-1GEA
42
GBP
APOSBIA0165-1G
APOSBIA0165-5G
APOSBIA0165-10G
Azithromycin
Azithromycin
MW:
749 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00873574 CAS Number: 83905-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...