4-Methoxybenzyl mercaptan 98%
Supplier: Apollo Scientific
Synonyms:
4-Methoxy-α-toluenethiol, p-Methoxytoluene-α-thiol
BI7572-5G
BI7572-25G
APOSBI7572-5GEA
91.8
GBP
APOSBI7572-5G
APOSBI7572-25G
4-Methoxybenzyl mercaptan 98%
4-Methoxybenzyl mercaptan
Formula:
C₈H₁₀OS MW: 154,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004871 CAS Number: 6258-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...