2-(4-Fluoro-3-methylphenyl)propan-2-ol

Supplier: Apollo Scientific

PC99017-1G PC99017-5G
APOSPC99017-1GEA 300 GBP
APOSPC99017-1G APOSPC99017-5G
2-(4-Fluoro-3-methylphenyl)propan-2-ol
2-(4-Fluoro-3-methylphenyl)propan-2-ol
Formula: C₁₀H₁₃FO
MW: 168,208 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17222275
CAS Number: 1267002-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR