(E)-1-Bromo-3,3,3-trifluoroprop-1-ene 95%
Supplier: Apollo Scientific
|
Warning
|
PC9761-1G
PC9761-5G
APOSPC9761-1GEA
211
GBP
APOSPC9761-1G
APOSPC9761-5G
(E)-1-Bromo-3,3,3-trifluoroprop-1-ene 95%
(E)-1-Bromo-3,3,3-trifluoroprop-1-ene
Formula:
C₃H₂BrF₃ MW: 174,947 g/mol Boiling Pt: 39…39,5 °C Melting Pt: 39…39,5 °C Density: >1,5 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01862034 CAS Number: 149597-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...