1,2,3-Trifluoropropane

Supplier: Apollo Scientific

PC9621-1G PC9621-5G
APOSPC9621-1GEA 125 GBP
APOSPC9621-1G APOSPC9621-5G
1,2,3-Trifluoropropane
1,2,3-Trifluoropropane
Formula: C₃H₅F₃
MW: 98,067 g/mol
Boiling Pt: 70…71 °C (760 mm Hg)
Density: 1,268 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD07784185
CAS Number: 66794-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR