2,3-Difluoro-4-(trifluoromethyl)phenylacetic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC9327-1G
PC9327-250MG
PC9327-5G
APOSPC9327-1GEA
317
GBP
APOSPC9327-1G
APOSPC9327-250MG
APOSPC9327-5G
2,3-Difluoro-4-(trifluoromethyl)phenylacetic acid 98%
2,3-Difluoro-4-(trifluoromethyl)phenylacetic acid
Formula:
C₉H₅F₅O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00236268 CAS Number: 247113-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...