3-(Trifluoromethyl)phenylacetyl chloride 97%

Supplier: Apollo Scientific
Danger

Synonyms: 3-(2-Chloro-2-oxoethyl)benzotrifluoride

PC9313-1G PC9313-5G PC9313-250MG
APOSPC9313-1GEA 77.3 GBP
APOSPC9313-1G APOSPC9313-5G APOSPC9313-250MG
3-(Trifluoromethyl)phenylacetyl chloride 97%
3-(Trifluoromethyl)phenylacetyl chloride
Formula: C₉H₆ClF₃O
MW: 222.592 g/mol
Boiling Pt: 94…96 °C (15 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08458137
CAS Number: 2003-14-7
UN: 1760
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR