4-Fluoro-1-isopropoxy-2-nitrobenzene 95%

Supplier: Apollo Scientific

PC911166-1G PC911166-5G
APOSPC911166-1GEA 211 GBP
APOSPC911166-1G APOSPC911166-5G
4-Fluoro-1-isopropoxy-2-nitrobenzene 95%
4-Fluoro-1-isopropoxy-2-nitrobenzene
Formula: C₉H₁₀FNO₃
MW: 199,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17014054
CAS Number: 1092496-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR