2-(3-Fluorophenylamino)acetic acid 95%
Supplier: Apollo Scientific
PC911148-1G
PC911148-250MG
APOSPC911148-1GEA
328
GBP
APOSPC911148-1G
APOSPC911148-250MG
2-(3-Fluorophenylamino)acetic acid 95%
2-(3-Fluorophenylamino)acetic acid
Formula:
C₈H₈FNO₂ MW: 169,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10029158 CAS Number: 5319-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...