3-(4-Fluorophenyl)-4H-1,2,4-triazole 98%

Supplier: Apollo Scientific

Synonyms: 3-(4-Fluorophenyl)-1H-1,2,4-triazole

PC910939-1G PC910939-5G
APOSPC910939-1GEA 145 GBP
APOSPC910939-1G APOSPC910939-5G
3-(4-Fluorophenyl)-4H-1,2,4-triazole 98%
3-(4-Fluorophenyl)-4H-1,2,4-triazole
Formula: C₈H₆FN₃
MW: 163,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18784761
CAS Number: 95728-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR