Ethyl 2-fluoropyridine-3-carboxylate 96%

Supplier: Apollo Scientific

Synonyms: Ethyl 2-fluoronicotinate

PC910913-1G PC910913-5G
APOSPC910913-1GEA 197 GBP
APOSPC910913-1G APOSPC910913-5G
Ethyl 2-fluoropyridine-3-carboxylate 96%
Ethyl 2-fluoropyridine-3-carboxylate
Formula: C₈H₈FNO₂
MW: 169,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698241
CAS Number: 113898-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR