2-Ethoxy-3-(trifluoromethyl)pyridine 98%

Supplier: Apollo Scientific

PC910720-1G PC910720-5G
APOSPC910720-1GEA 223 GBP
APOSPC910720-1G APOSPC910720-5G
2-Ethoxy-3-(trifluoromethyl)pyridine 98%
2-Ethoxy-3-(trifluoromethyl)pyridine
Formula: C₈H₈F₃NO
MW: 191,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044343
CAS Number: 849934-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR