2-(Trifluoromethyl)-4H-1-benzopyran-4-one ≥98%

Supplier: Apollo Scientific

PC910374-100MG PC910374-1G PC910374-5G
APOSPC910374-100MGEA 19.4 GBP
APOSPC910374-100MG APOSPC910374-1G APOSPC910374-5G
2-(Trifluoromethyl)-4H-1-benzopyran-4-one ≥98%
2-(Trifluoromethyl)-4H-1-benzopyran-4-one
Formula: C₁₀H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00761957
CAS Number: 151668-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR