2,3,4,6-Tetrafluorobenzoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC9098-1G
PC9098-250MG
APOSPC9098-1GEA
109
GBP
APOSPC9098-1G
APOSPC9098-250MG
2,3,4,6-Tetrafluorobenzoic acid 98%
2,3,4,6-Tetrafluorobenzoic acid
Formula:
C₇H₂F₄O₂ MW: 194,083 g/mol Melting Pt: 97…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039523 CAS Number: 32890-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...