3-(3-Fluorophenoxy)propylamine 95%
Supplier: Apollo Scientific
PC908667-1G
PC908667-5G
APOSPC908667-1GEA
184
GBP
APOSPC908667-1G
APOSPC908667-5G
3-(3-Fluorophenoxy)propylamine 95%
3-(3-Fluorophenoxy)propylamine
Formula:
C₉H₁₂FNO MW: 169.196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09741966 CAS Number: 116753-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...