2,6-Difluoro-3,5-dimethoxyaniline 95%

Supplier: Apollo Scientific

PC908567-1G PC908567-250MG
APOSPC908567-1GEA 160 GBP
APOSPC908567-1G APOSPC908567-250MG
2,6-Difluoro-3,5-dimethoxyaniline 95%
2,6-Difluoro-3,5-dimethoxyaniline
Formula: C₈H₉F₂NO₂
MW: 189,159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12973804
CAS Number: 651734-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR