6-Chloro-5-fluoro-2,3-dihydro-1H-inden-1-one 95+%

Supplier: Apollo Scientific
Warning

PC908169-100MG PC908169-250MG PC908169-1G
APOSPC908169-100MGEA 91.8 GBP
APOSPC908169-100MG APOSPC908169-250MG APOSPC908169-1G
6-Chloro-5-fluoro-2,3-dihydro-1H-inden-1-one 95+%
6-Chloro-5-fluoro-2,3-dihydro-1H-inden-1-one
Formula: C₉H₆ClFO
MW: 184,595 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12547664
CAS Number: 881189-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR