4-Fluoro-1,2-benzoxazol-3-amine 97%

Supplier: Apollo Scientific

PC907849-1G PC907849-5G
APOSPC907849-1GEA 197 GBP
APOSPC907849-1G APOSPC907849-5G
4-Fluoro-1,2-benzoxazol-3-amine 97%
4-Fluoro-1,2-benzoxazol-3-amine
Formula: C₇H₅FN₂O
MW: 152,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09033911
CAS Number: 904815-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR