1-Difluoromethyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid 95%
Supplier: Apollo Scientific
PC907410-1G
PC907410-250MG
APOSPC907410-1GEA
728
GBP
APOSPC907410-1G
APOSPC907410-250MG
1-Difluoromethyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid 95%
1-Difluoromethyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid
Formula:
C₇H₅F₂NO₃ MW: 189,116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18791579 CAS Number: 1129458-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...