Methyl 3-oxo-4-(2,4,5-trifluorophenyl)butanoate 97%

Supplier: Apollo Scientific

PC905447-10G PC905447-1G PC905447-5G
APOSPC905447-10GEA 205 GBP
APOSPC905447-10G APOSPC905447-1G APOSPC905447-5G
Methyl 3-oxo-4-(2,4,5-trifluorophenyl)butanoate 97%
Methyl 3-oxo-4-(2,4,5-trifluorophenyl)butanoate
Formula: C₁₁H₉F₃O₃
MW: 246,183 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09833197
CAS Number: 769195-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR