5-(4-Fluorophenyl)isoxazol-3-amine 95%

Supplier: Apollo Scientific

PC905317-1G PC905317-5G
APOSPC905317-1GEA 112 GBP
APOSPC905317-1G APOSPC905317-5G
5-(4-Fluorophenyl)isoxazol-3-amine 95%
5-(4-Fluorophenyl)isoxazol-3-amine
Formula: C₉H₇FN₂O
MW: 178,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08447045
CAS Number: 925005-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR