4-(2,2,2-Trifluoroacetamido)butanoic acid 95%

Supplier: Apollo Scientific

PC905290-1G PC905290-250MG
APOSPC905290-1GEA 73.3 GBP
APOSPC905290-1G APOSPC905290-250MG
4-(2,2,2-Trifluoroacetamido)butanoic acid 95%
4-(2,2,2-Trifluoroacetamido)butanoic acid
Formula: C₆H₈F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00130210
CAS Number: 50632-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR