2-Chloro-5'-fluoroacetophenone 95%

Supplier: Apollo Scientific

Synonyms: 2-Chloro-1-(3-fluorophenyl)ethanone

PC905062-1G PC905062-250MG
APOSPC905062-1GEA 476 GBP
APOSPC905062-1G APOSPC905062-250MG
2-Chloro-5'-fluoroacetophenone 95%
2-Chloro-5'-fluoroacetophenone
Formula: C₈H₆ClFO
Melting Pt: 27…32 °C
Storage Temperature: Ambient
MDL Number: MFCD09992878
CAS Number: 53688-18-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR