3-(6-Fluoro-pyridin-2-ylamino)-propan-1-ol 95%

Supplier: Apollo Scientific

PC904985-1G PC904985-250MG
APOSPC904985-1GEA 394 GBP
APOSPC904985-1G APOSPC904985-250MG
3-(6-Fluoro-pyridin-2-ylamino)-propan-1-ol 95%
3-(6-Fluoro-pyridin-2-ylamino)-propan-1-ol
Formula: C₈H₁₁FN₂O
MW: 170,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14600210
CAS Number: 1000981-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR