2-[(Ethylamino)methyl]-4-fluorophenol 95%
Supplier: Apollo Scientific
PC904176-1G
PC904176-250MG
APOSPC904176-1GEA
394
GBP
APOSPC904176-1G
APOSPC904176-250MG
2-[(Ethylamino)methyl]-4-fluorophenol 95%
2-[(Ethylamino)methyl]-4-fluorophenol
Formula:
C₉H₁₂FNO MW: 169,196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21641326 CAS Number: 1363166-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...