2-Fluoro-4-methoxycinnamic acid 95%

Supplier: Apollo Scientific

PC902714-1G PC902714-250MG
APOSPC902714-1GEA 296 GBP
APOSPC902714-1G APOSPC902714-250MG
2-Fluoro-4-methoxycinnamic acid 95%
2-Fluoro-4-methoxycinnamic acid
Formula: C₁₀H₉FO₃
MW: 196,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03002812
CAS Number: 696589-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR