1-[3-(Trifluoromethyl)pyrazol-1-yl]propan-2-ol 95%

Supplier: Apollo Scientific

PC902235-1G PC902235-5G
APOSPC902235-1GEA 243 GBP
APOSPC902235-1G APOSPC902235-5G
1-[3-(Trifluoromethyl)pyrazol-1-yl]propan-2-ol 95%
1-[3-(Trifluoromethyl)pyrazol-1-yl]propan-2-ol
Formula: C₇H₉F₃N₂O
MW: 194,154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16065387
CAS Number: 742096-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR