Methyl 2-(4-fluorophenyl)-2-methylpropanoate 98%
Supplier: Apollo Scientific
PC902101-1G
PC902101-5G
APOSPC902101-1GEA
185
GBP
APOSPC902101-1G
APOSPC902101-5G
Methyl 2-(4-fluorophenyl)-2-methylpropanoate 98%
Methyl 2-(4-fluorophenyl)-2-methylpropanoate
Formula:
C₁₁H₁₃FO₂ MW: 196,218 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23096917 CAS Number: 476429-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...