1-Ethyl-5-methyl-3-(trifluoromethyl)pyrazole 95%

Supplier: Apollo Scientific

PC902079-10G PC902079-1G PC902079-5G
APOSPC902079-10GEA 683 GBP
APOSPC902079-10G APOSPC902079-1G APOSPC902079-5G
1-Ethyl-5-methyl-3-(trifluoromethyl)pyrazole 95%
1-Ethyl-5-methyl-3-(trifluoromethyl)pyrazole
Formula: C₇H₉F₃N₂
MW: 178,155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25121754
CAS Number: 128694-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR