4-Fluoro-2-isopropoxy-1-nitrobenzene 98%

Supplier: Apollo Scientific

PC901654-10G PC901654-1G PC901654-5G
APOSPC901654-10GEA 779 GBP
APOSPC901654-10G APOSPC901654-1G APOSPC901654-5G
4-Fluoro-2-isopropoxy-1-nitrobenzene 98%
4-Fluoro-2-isopropoxy-1-nitrobenzene
Formula: C₉H₁₀FNO₃
MW: 199.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115631
CAS Number: 28987-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR