Methyl 3-(2-fluorophenyl)propanoate 97%

Supplier: Apollo Scientific

PC901544-1G PC901544-5G
APOSPC901544-1GEA 72.2 GBP
APOSPC901544-1G APOSPC901544-5G
Methyl 3-(2-fluorophenyl)propanoate 97%
Methyl 3-(2-fluorophenyl)propanoate
Formula: C₁₀H₁₁FO₂
MW: 182,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031751
CAS Number: 143654-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR