3-(3-Bromo-2-fluorophenyl)propanoic acid 95%

Supplier: Apollo Scientific

PC901542-100MG PC901542-1G PC901542-250MG
APOSPC901542-100MGEA 19.4 GBP
APOSPC901542-100MG APOSPC901542-1G APOSPC901542-250MG
3-(3-Bromo-2-fluorophenyl)propanoic acid 95%
3-(3-Bromo-2-fluorophenyl)propanoic acid
Formula: C₉H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD18391365
CAS Number: 1261814-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR