4,4-Difluorocyclohexan-1-amine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4,4-Difluorocyclohexanamine
PC901499-1G
PC901499-250MG
APOSPC901499-1GEA
257
GBP
APOSPC901499-1G
APOSPC901499-250MG
4,4-Difluorocyclohexan-1-amine 97%
4,4-Difluorocyclohexan-1-amine
Formula:
C₆H₁₁F₂N MW: 135,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06409984 CAS Number: 458566-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...